This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM161A SIRNA Assays ›
Gene Symbol:
Tmem161a
Gene Name:
transmembrane protein 161A
Gene Aliases:
AI428876, BB161850, BC021367
Chromosome Location:
Chr.8: 70172329 - 70183681 on Build GRCm38
UniGene ID:
Mm.23488
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s107814

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem161a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293797.1 NP_001280726.1 5 401
NM_145597.4 NP_663572.1 6 505
GenBank mRNA AK049979.1 - 6 504
AK082758.1 - 6 501
AK082916.1 - 5 402
AK088569.1 - 5 391
AK147962.1 - 6 498
BC021367.1 - 6 505 5030434

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to retinoic acid
cellular response to oxidative stress
cellular response to UV
positive regulation of DNA repair
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0