This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRAFD1 SIRNA Assays ›
Gene Symbol:
Trafd1
Gene Name:
TRAF type zinc finger domain containing 1
Gene Aliases:
1110008K06Rik, Fln29
Chromosome Location:
Chr.5: 121371725 - 121386480 on Build GRCm38
UniGene ID:
Mm.20488
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s106937

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Trafd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163470.1 NP_001156942.1 5 426
NM_172275.2 NP_758479.2 5 426
GenBank mRNA AK150038.1 - 5 395
BC038396.1 - 5 382 3987227
BU613615.1 - 5 426

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_053760, XM_008769209, XM_006249376.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of innate immune response
protein binding
zinc ion binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0