This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LEPROT SIRNA Assays ›
Gene Symbol:
Leprot
Gene Name:
leptin receptor overlapping transcript
Gene Aliases:
LEPR, OB-RGRP, Obrgrp, VPS55
Chromosome Location:
Chr.4: 101647783 - 101659358 on Build GRCm38
UniGene ID:
Mm.4756
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s106470

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Leprot
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175036.4 NP_778201.1 3 289
GenBank mRNA AJ011565.1 - 3 262
AK009569.1 - 3 290
AK088848.1 - 1 106
AK152008.1 - 3 356
AK159489.1 - 3 355
AK160453.1 - 3 303
AK165821.1 - 3 348
AK168597.1 - 3 325
BC004744.1 - 3 300 3583760
BC010289.1 - 3 210 2651399

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of JAK-STAT cascade
negative regulation of growth hormone receptor signaling pathway
positive regulation of protein targeting to mitochondrion
negative regulation of protein localization to cell surface
receptor binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0