This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRMT13 SIRNA Assays ›
Gene Symbol:
Trmt13
Gene Name:
tRNA methyltransferase 13
Gene Aliases:
4631408H19Rik, A330067P21, A930028L21Rik, Ccdc76
Chromosome Location:
Chr.3: 116581093 - 116614587 on Build GRCm38
UniGene ID:
Mm.259648
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s106316

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Trmt13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030016.2 NP_084292.1 4 1853
GenBank mRNA AK039588.1 - 4 1854
AK041636.1 - 3 319
AK163561.1 - 3 268
BC127174.1 - 3 211

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001033902, BC103656, BC166503.

Gene Ontology Categories:

Function(s) Process(es)

tRNA processing
biological_process
tRNA methylation
methylation
methyltransferase activity
tRNA methyltransferase activity
transferase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0