This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SMOX SIRNA Assays ›
Gene Symbol:
Smox
Gene Name:
spermine oxidase
Gene Aliases:
B130066H01Rik, PAO, PAOh1, SMO
Chromosome Location:
Chr.2: 131491782 - 131525183 on Build GRCm38
UniGene ID:
Mm.136586
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s105946

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Smox
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177833.1 NP_001171304.1 3 541
NM_001177834.1 NP_001171305.1 3 541
NM_001177835.1 NP_001171306.1 3 541
NM_001177836.1 NP_001171307.1 3 541
NM_001177837.1 NP_001171308.1 3 541
NM_001177838.1 NP_001171309.1 3 541
NM_001177839.1 NP_001171310.1 3 541
NM_001177840.1 NP_001171311.1 3 541
NM_145533.2 NP_663508.1 3 541
GenBank mRNA AF495851.1 - 3 409
AF495852.1 - 3 410
AF495853.1 - 3 410
AJ567473.1 - 3 460
AJ567474.1 - 3 460
AJ567475.1 - 3 460
AJ567476.1 - 3 460
AJ567477.1 - 3 460
AJ567478.1 - 3 460
AJ567479.1 - 3 460
AJ567480.1 - 3 460
AK087437.1 - 1 1487
AK143129.1 - 3 515
AK159275.1 - 3 468
AK159372.1 - 3 468
AK166350.1 - 440
BC004831.1 - 3 460 2647695

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

polyamine catabolic process
spermine catabolic process
oxidation-reduction process
oxidoreductase activity
polyamine oxidase activity
norspermine:oxygen oxidoreductase activity
N1-acetylspermine:oxygen oxidoreductase (N1-acetylspermidine-forming) activity
spermine:oxygen oxidoreductase (spermidine-forming) activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0