This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FRMD5 SIRNA Assays ›
Gene Symbol:
Frmd5
Gene Name:
FERM domain containing 5
Gene Aliases:
1500032A09Rik, 5830442F04, A930004K21Rik, R75430
Chromosome Location:
Chr.2: 121545529 - 121807757 on Build GRCm38
UniGene ID:
Mm.121514
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s105928

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Frmd5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172673.3 NP_766261.2 6, 7 777
GenBank mRNA AK030883.1 - 6, 7 533
AK044270.1 - 6, 7 528
AK048865.1 - 6, 7 778
AK054484.1 - 6, 7 734
BC038623.1 - 6, 7 514
BC057549.1 - 3, 4 306 6416914
BC062889.1 - 6, 7 734 5698263

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008762214, XM_008762211, XM_008762213, XM_008762212, XM_008762215.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
cytoskeletal protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0