This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AMBRA1 SIRNA Assays ›
Gene Symbol:
Ambra1
Gene Name:
autophagy/beclin 1 regulator 1
Gene Aliases:
2310079H06Rik, A130023A14, AA474864, AV021921, D030051N19Rik, mKIAA1736
Chromosome Location:
Chr.2: 91730111 - 91918849 on Build GRCm38
UniGene ID:
Mm.436667
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s105886

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ambra1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080754.1 NP_001074223.1 6, 7 987
NM_172669.3 NP_766257.3 6, 7 987
GenBank mRNA AK037516.1 - 6, 7 944
AK083596.1 - 6, 7 944
AK084557.1 - 6, 7 969
AK129433.1 - 2, 3 251
AK148887.1 - 6, 7 979
AK155513.1 - 6, 7 950
AK169280.1 - 6, 7 940
BC032293.1 - 6, 7 910
BC054082.1 - 6, 7 919

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

autophagy
multicellular organism development
nervous system development
negative regulation of cell proliferation
embryo development
neural tube development
cell differentiation
negative regulation of neuron apoptotic process
positive regulation of phosphatidylinositol 3-kinase activity
mitophagy in response to mitochondrial depolarization
positive regulation of mitophagy
regulation of autophagosome assembly
protein binding
ubiquitin protein ligase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0