This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MAPKAP1 SIRNA Assays ›
Gene Symbol:
Mapkap1
Gene Name:
mitogen-activated protein kinase associated protein 1
Gene Aliases:
AI591529, D230039K05Rik, Sin1
Chromosome Location:
Chr.2: 34406771 - 34624958 on Build GRCm38
UniGene ID:
Mm.270866
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s105796

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mapkap1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290625.1 NP_001277554.1 10 1414
NM_001290626.1 NP_001277555.1 9 1412
NM_177345.4 NP_796319.1 10 1740
GenBank mRNA AK027932.1 - 5 513
AK042154.1 - 9 1402
AK052045.1 - 10 1408
AK132263.1 - 10 1404
AK148428.1 - 10 1730
BC027377.1 - 5 542
BC031579.1 - 5 542
BC043296.1 - 10 1639
BC090644.1 - 9 1314 30629263
BC096618.1 - 10 1713 6392317

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_024117, NM_001006619, NM_001006617, NM_001006621, NM_001006620, and others.
This siRNA also targets the following transcripts in Rat: NM_001011964, XM_006233950, XM_008761722, XM_008761720, XM_008761723, and others.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of Ras protein signal transduction
protein binding
phosphatidylinositol-4,5-bisphosphate binding
phosphatidylinositol-3,4,5-trisphosphate binding
Ras GTPase binding
protein kinase binding
phosphatidylinositol-3,4-bisphosphate binding
phosphatidic acid binding
phosphatidylinositol-3,5-bisphosphate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0