This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NDUFS1 SIRNA Assays ›
Gene Symbol:
Ndufs1
Gene Name:
NADH dehydrogenase (ubiquinone) Fe-S protein 1
Gene Aliases:
5830412M15Rik, 9930026A05Rik
Chromosome Location:
Chr.1: 63143592 - 63176822 on Build GRCm38
UniGene ID:
Mm.290791
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s105592

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ndufs1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160038.1 NP_001153510.1 11 1425
NM_001160039.1 NP_001153511.1 11 1267
NM_001160040.1 NP_001153512.1 10 1321
NM_145518.2 NP_663493.2 10 1163
GenBank mRNA AK034597.1 - 10 1232
AK036926.1 - 10 1028
AK078590.1 - 10 1010
AK142711.1 - 10 1043
AK146814.1 - 10 1181
AK166604.1 - 11 1114
AK167705.1 - 10 1011
AK202692.1 - 27
BC006660.1 - 10 1050 3501043
BC015300.1 - 10 983 4236882

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
apoptotic mitochondrial changes
ATP synthesis coupled electron transport
cellular respiration
ATP metabolic process
regulation of mitochondrial membrane potential
oxidation-reduction process
reactive oxygen species metabolic process
NADH dehydrogenase activity
NADH dehydrogenase (ubiquinone) activity
electron carrier activity
oxidoreductase activity
oxidoreductase activity, acting on NAD(P)H
metal ion binding
iron-sulfur cluster binding
2 iron, 2 sulfur cluster binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0