This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CD226 SIRNA Assays ›
Gene Symbol:
Cd226
Gene Name:
CD226 antigen
Gene Aliases:
BC051526, DNAM-1, DNAM1, Pta1, TLiSA1
Chromosome Location:
Chr.18: 89176954 - 89272232 on Build GRCm38
UniGene ID:
Mm.139293
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s105184

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cd226
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039149.1 NP_001034238.1 3 392
NM_178687.2 NP_848802.2 4 731
GenBank mRNA AB195429.1 - 3 504
AF416980.1 - 4 563
AF421198.1 - 4 563
AF421199.1 - 3 224
AF421200.1 - 2 152
AK036455.1 - 5 826
AK140349.1 - 4 619
BC051526.1 - 3 326
BC132422.1 - 4 676 40130699
BC138769.1 - 4 676 8861791
BC145410.1 - 3 337 9053968
BC145411.1 - 4 676

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cytokine production
positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
positive regulation of immunoglobulin mediated immune response
cell adhesion
positive regulation of mast cell activation
positive regulation of natural killer cell mediated cytotoxicity
positive regulation of Fc receptor mediated stimulatory signaling pathway
integrin binding
protein binding
protein kinase binding
cell adhesion molecule binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0