This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KRT79 SIRNA Assays ›
Gene Symbol:
Krt79
Gene Name:
keratin 79
Gene Aliases:
BC031593
Chromosome Location:
Chr.15: 101929332 - 101940324 on Build GRCm38
UniGene ID:
Mm.162744
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s104668

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Krt79
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146063.1 NP_666175.1 1, 2 571
GenBank mRNA BC031593.1 - 1, 2 571 4935395

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_003750410.

Gene Ontology Categories:

Process(es)

structural molecule activity
enzyme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7bff9b55f5-ncxmj:80/100.66.128.142:80.
git-commit: 5f36da1a8ad9bc9722227bfd86ebc827ae27d083
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0