This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SENP1 SIRNA Assays ›
Gene Symbol:
Senp1
Gene Name:
SUMO1/sentrin specific peptidase 1
Gene Aliases:
2310046A20Rik, D15Ertd528e, E330036L07Rik, suPr-2
Chromosome Location:
Chr.15: 98038744 - 98093569 on Build GRCm38
UniGene ID:
Mm.384023
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s104660

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Senp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144851.5 NP_659100.1 13 1673
GenBank mRNA AK028565.1 - 13 1415
AK028762.1 - 5 2647
AK035723.1 - 14 1856
AK054521.1 - 14 1805
AK088597.1 - 13 1417
BC023129.1 - 12 1387 3595534

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
activation of cysteine-type endopeptidase activity involved in apoptotic process
multicellular organism development
regulation of definitive erythrocyte differentiation
protein desumoylation
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
positive regulation of transcription from RNA polymerase II promoter
apoptotic signaling pathway
protein binding
peptidase activity
cysteine-type peptidase activity
hydrolase activity
SUMO-specific protease activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0