This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ANKRD54 SIRNA Assays ›
Gene Symbol:
Ankrd54
Gene Name:
ankyrin repeat domain 54
Gene Aliases:
C730048E16Rik, EST1068184, EST475269, Liar
Chromosome Location:
Chr.15: 79053094 - 79062958 on Build GRCm38
UniGene ID:
Mm.24262
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s104578

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ankrd54
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144849.1 NP_659098.1 7 866
GenBank mRNA AK050440.1 - 6 1581
AK133785.1 - 7 890
AK143625.1 - 7 893
BC014743.1 - 7 866 4503319
DQ884878.1 - 7 894

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001025285, XM_006242012, BC093616.

Gene Ontology Categories:

Function(s) Process(es)

nucleocytoplasmic transport
positive regulation of erythrocyte differentiation
regulation of protein kinase activity
regulation of intracellular signal transduction
protein binding
protein kinase regulator activity
protein complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0