This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other WNT9A SIRNA Assays ›
Gene Symbol:
Wnt9a
Gene Name:
wingless-type MMTV integration site family, member 9A
Gene Aliases:
Wnt14
Chromosome Location:
Chr.11: 59306849 - 59333552 on Build GRCm38
UniGene ID:
Mm.218794
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s103705

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Wnt9a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139298.2 NP_647459.1 4 908
GenBank mRNA AB072311.1 - 4 872
AK087490.1 - 4 915
BC066165.1 - 4 885 30435371

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitotic cell cycle checkpoint
multicellular organism development
negative regulation of cell proliferation
Wnt signaling pathway
neuron differentiation
negative regulation of chondrocyte differentiation
embryonic forelimb morphogenesis
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
cell fate commitment
positive regulation of cell differentiation
positive regulation of smoothened signaling pathway
embryonic skeletal system morphogenesis
anatomical structure development
canonical Wnt signaling pathway
negative regulation of cell death
negative regulation of cartilage development
receptor binding
frizzled binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0