This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CFP SIRNA Assays ›
Gene Symbol:
CFP
Gene Name:
complement factor properdin
Gene Aliases:
BFD, PFC, PFD, PROPERDIN
Chromosome Location:
Chr.X: 47624213 - 47630305 on Build GRCh38
UniGene ID:
Hs.53155
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s10334

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CFP
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145252.1 NP_001138724.1 8 1272
NM_002621.2 NP_002612.1 9 1388
XM_017029575.1 XP_016885064.1 7 1894
GenBank mRNA AK122955.1 - 7 2444
AK310695.1 - 8 1267
BC015756.1 - 8 1206 4850121
KU178249.1 - 8 1146
KU178250.1 - 9 1076
M83652.1 - 8 1218
X57748.1 - 8 1211

Back To Top

More Information


Gene Ontology Categories:

Function(s)

immune response
complement activation
complement activation, alternative pathway
regulation of complement activation
protein O-linked fucosylation
defense response to bacterium

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header