This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DIS3L SIRNA Assays ›
Gene Symbol:
Dis3l
Gene Name:
DIS3 mitotic control homolog (S. cerevisiae)-like
Gene Aliases:
AV340375
Chromosome Location:
Chr.9: 64306756 - 64341423 on Build GRCm38
UniGene ID:
Mm.268341
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s102910

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dis3l
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001295.2 NP_001001295.2 10 1609
NM_001177784.1 NP_001171255.1 10 1570
NM_172519.3 NP_766107.1 10 1513
GenBank mRNA AK029974.1 - 10 1514
AK032451.1 - 10 1518
AK077031.1 - 10 1573
AK150851.1 - 10 1559
AK151780.1 - 10 1559
AK153433.1 - 10 1559
AK173310.1 - 10 1543
BC056939.1 - 10 1542 5708685

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001008380, XM_008766311, XM_006243262, XM_006243261, XM_006243263, and others.

Gene Ontology Categories:

Function(s) Process(es)

rRNA processing
mRNA catabolic process
rRNA catabolic process
RNA phosphodiester bond hydrolysis, exonucleolytic
3'-5'-exoribonuclease activity
RNA binding
nuclease activity
exonuclease activity
5'-3' exoribonuclease activity
hydrolase activity
enzyme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0