This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GABPB2 SIRNA Assays ›
Gene Symbol:
Gabpb2
Gene Name:
GA repeat binding protein, beta 2
Gene Aliases:
1810015F01Rik, 5830427M07, 9430006E19Rik, A430024B14Rik, AV050852, Gabpb2-1
Chromosome Location:
Chr.3: 95181766 - 95217951 on Build GRCm38
UniGene ID:
Mm.224820
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s102781

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gabpb2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029885.1 NP_084161.1 9 1377
NM_172512.2 NP_766100.1 8 1280
GenBank mRNA AK030849.1 - 8 1049
AK036692.1 - 8 1861
AK137604.1 - 1 2646
AK141382.1 - 9 1351
AK163329.1 - 2 216
BC150160.1 - 8 942 40141577
BC150482.1 - 8 942 40141578

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
protein homodimerization activity
transcription regulatory region DNA binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0