This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MFSD9 SIRNA Assays ›
Gene Symbol:
Mfsd9
Gene Name:
major facilitator superfamily domain containing 9
Gene Aliases:
4931419K03Rik
Chromosome Location:
Chr.1: 40772040 - 40790657 on Build GRCm38
UniGene ID:
Mm.235443
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s102507

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mfsd9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172499.2 NP_766087.1 6 922
GenBank mRNA AK029874.1 - 6 923
AK169911.1 - 7 1083
BC057633.1 - 6 928 4035998

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_032718, BC006242, AK075376, BC009310, AK296768, and others.
This siRNA also targets the following transcripts in Rat: NM_001108215, XM_006244823, XM_006244824.

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
transmembrane transport
molecular_function
transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0