This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZDHHC9 SIRNA Assays ›
Gene Symbol:
Zdhhc9
Gene Name:
zinc finger, DHHC domain containing 9
Gene Aliases:
6430508G22, 9530098M12Rik
Chromosome Location:
Chr.X: 48171967 - 48208702 on Build GRCm38
UniGene ID:
Mm.207367
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s101961

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zdhhc9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172465.4 NP_766053.1 4 719
GenBank mRNA AK032233.1 - 4 701
AK035745.1 - 3 2142
AK079248.1 - 4 686
BC042618.1 - 4 578 4950217
BC090832.1 - 4 537 6401763

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001039016, XM_006257521, BC129070, AY886526.

Gene Ontology Categories:

Function(s) Process(es)

peptidyl-L-cysteine S-palmitoylation
protein palmitoylation
zinc ion binding
palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
protein-cysteine S-palmitoyltransferase activity
Ras palmitoyltransferase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0