This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PHLDB2 SIRNA Assays ›
Gene Symbol:
Phldb2
Gene Name:
pleckstrin homology-like domain, family B, member 2
Gene Aliases:
AV253284, C820004H04Rik, LL5b, LL5beta
Chromosome Location:
Chr.16: 45746230 - 45953701 on Build GRCm38
UniGene ID:
Mm.211477
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s101808

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Phldb2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252442.1 NP_001239371.1 2 719
NM_153412.4 NP_700461.2 2 719
GenBank mRNA AF506821.1 - 2 720
AK049595.1 - 2 573
AK050493.1 - 3 886
AK165251.1 - 2 681
AK199939.1 - 1 181
BC050915.1 - 2 575 6405379
BC060683.1 - 2 575 6405379

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0