This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HPS4 SIRNA Assays ›
Gene Symbol:
Hps4
Gene Name:
Hermansky-Pudlak syndrome 4 homolog (human)
Gene Aliases:
2010205O06Rik, AU040608, BLOC-3, C130020P05Rik, le, mKIAA1667
Chromosome Location:
Chr.5: 112343056 - 112378424 on Build GRCm38
UniGene ID:
Mm.238043
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s101319

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hps4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138646.3 NP_619587.3 11 1604
GenBank mRNA AK081489.1 - 10 1958
AK129418.1 - 10 1491
AK147954.1 - 10 2047
AK171871.1 - 11 1573
AY043415.1 - 10 1124
BC004668.1 - 1 211
BC063055.1 - 10 1296 5701206

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein targeting
organelle organization
lysosome organization
blood coagulation
melanocyte differentiation
protein stabilization
positive regulation of protein targeting to mitochondrion
protein homodimerization activity
protein dimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm