This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SUGCT SIRNA Assays ›
Gene Symbol:
Sugct
Gene Name:
succinyl-CoA glutarate-CoA transferase
Gene Aliases:
5033411D12Rik, D17907
Chromosome Location:
Chr.13: 16857472 - 17694793 on Build GRCm38
UniGene ID:
Mm.364180
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s101214

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sugct
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138654.3 NP_619595.3 5 379
GenBank mRNA AF397014.1 - 5 395
AK160265.1 - 4 252
BC055852.1 - 5 353 4221852
CJ068463.1 - 5 381

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001014146, BC079348.

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
catalytic activity
transferase activity
succinate-hydroxymethylglutarate CoA-transferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0