This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC26A6 SIRNA Assays ›
Gene Symbol:
Slc26a6
Gene Name:
solute carrier family 26, member 6
Gene Aliases:
B930010B04Rik, CFEX, Pat-1, Pat1
Chromosome Location:
Chr.9: 108854043 - 108862143 on Build GRCm38
UniGene ID:
Mm.45201
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s101167

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc26a6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134420.4 NP_599252.2 2059
GenBank mRNA AB099881.1 - 1220
AF248494.1 - 1405
AK029238.1 - 2981
AK046993.1 - 3319
AK053088.1 - 1710
AK080750.1 - 1364
AK163670.1 - 2060
AK163671.1 - 2060
AY032863.1 - 1364
AY049076.1 - 1319
BC028856.1 - 1383 4165725

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
anion transport
chloride transport
sulfate transport
bicarbonate transport
formate transport
mannitol transport
oxalate transport
transepithelial chloride transport
angiotensin-activated signaling pathway
epithelial fluid transport
oxalic acid secretion
sperm capacitation
intestinal absorption
regulation of intracellular pH
intracellular pH elevation
transmembrane transport
protein kinase C signaling
transepithelial transport
cellular response to cAMP
cellular response to fructose stimulus
cellular response to interferon-gamma
anion transmembrane transport
sulfate transmembrane transport
chloride transmembrane transport
organic acid transmembrane transport
positive regulation of dipeptide transmembrane transport
chloride channel activity
protein binding
secondary active sulfate transmembrane transporter activity
anion transmembrane transporter activity
bicarbonate transmembrane transporter activity
chloride transmembrane transporter activity
sulfate transmembrane transporter activity
antiporter activity
anion:anion antiporter activity
formate transmembrane transporter activity
efflux transmembrane transporter activity
formate uptake transmembrane transporter activity
formate efflux transmembrane transporter activity
oxalate transmembrane transporter activity
PDZ domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0