siRNA Details
Gene Symbol: Vmn1r235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134199.3 | NP_598960.3 | 2 | 929 | ||
GenBank mRNA | AF394953.1 | - | 2 | 930 |

Gene Symbol: Vmn1r235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134199.3 | NP_598960.3 | 2 | 929 | ||
GenBank mRNA | AF394953.1 | - | 2 | 930 |