This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MCOLN3 SIRNA Assays ›
Gene Symbol:
Mcoln3
Gene Name:
mucolipin 3
Gene Aliases:
6720490O21Rik, TRPML3, Va
Chromosome Location:
Chr.3: 146093277 - 146141808 on Build GRCm38
UniGene ID:
Mm.114683
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s100874

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mcoln3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134160.1 NP_598921.1 8 1029
GenBank mRNA AF475086.1 - 8 1029
AK030819.1 - 8 1149
AK033008.1 - 8 1143
AK035029.1 - 8 1150
AY083531.1 - 8 1027
BC106856.2 - 8 1029 40047039
BC106857.2 - 8 1029 40047042

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001012059, BC079200.

Gene Ontology Categories:

Function(s)

transport
ion transport
sensory perception of sound
locomotory behavior
auditory receptor cell differentiation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0