This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RFXAP SIRNA Assays ›
Gene Symbol:
Rfxap
Gene Name:
regulatory factor X-associated protein
Gene Aliases:
5730495K23Rik
Chromosome Location:
Chr.3: 54803115 - 54807791 on Build GRCm38
UniGene ID:
Mm.185735
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s100775

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rfxap
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133231.2 NP_573494.1 1, 2 601
GenBank mRNA AF335512.1 - 1, 2 581
AK077640.1 - 1, 2 599
AK087489.1 - 1, 2 602
AK144633.1 - 1, 2 588
BC019935.1 - 1, 2 564 5041039
BC047123.1 - 1, 2 564

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of transcription, DNA-templated
DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0