This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP287 SIRNA Assays ›
Gene Symbol:
Zfp287
Gene Name:
zinc finger protein 287
Gene Aliases:
B230333C16Rik, SKAT-2, Skat2, Znf287, mszf16, mszf74
Chromosome Location:
Chr.11: 62692444 - 62731900 on Build GRCm38
UniGene ID:
Mm.339982
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s100711

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp287
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133208.2 NP_573471.2 5 2544
GenBank mRNA AF281141.1 - 5 2544
AK029572.1 - 5 2124
BC023790.1 - 3 1563
BC117876.1 - 5 2269 40095930
BC117877.1 - 5 2269 40095933

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of cytokine biosynthetic process
positive regulation of transcription, DNA-templated
nucleic acid binding
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0