This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EHBP1L1 SIRNA Assays ›
Gene Symbol:
Ehbp1l1
Gene Name:
EH domain binding protein 1-like 1
Gene Aliases:
G430002G23Rik
Chromosome Location:
Chr.19: 5707369 - 5726431 on Build GRCm38
UniGene ID:
Mm.210447
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s100133

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ehbp1l1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114595.1 NP_001108067.1 15 2245
NM_001114596.1 NP_001108068.1 16 2281
NM_001114597.1 NP_001108069.1 16 5113
NM_053252.3 NP_444482.2 16 2590
GenBank mRNA AF305087.1 - 16 5060
AF305088.1 - 16 2534
AF305089.1 - 15 2189
AF305090.1 - 16 2225
AK087817.1 - 12 1832
AK089917.1 - 15 2199
BC004660.1 - 16 2530 3497913

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001270704, NM_001129997, NM_001025117, NM_001270705, XM_008760139, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0