siRNA Details
Gene Symbol: LOC729867 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123725.1 | 3 | 495 | |||
GenBank mRNA | AK093299.1 | - | 3 | 495 |

Gene Symbol: LOC729867 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123725.1 | 3 | 495 | |||
GenBank mRNA | AK093299.1 | - | 3 | 495 |