siRNA Details
Gene Symbol: LOC101927379 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109881.1 | 1 | 43 | |||
GenBank mRNA | BC043373.1 | - | 1 | 42 | ||
BX119826.1 | - | 1 | 168 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI832297.1 | - | 1 | 50 | Chr. 5 - 122704273 - 122730556 [-] on Build GRCh38 |
