siRNA Details
Gene Symbol: LOC100506271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110515.1 | 1 | 399 | |||
GenBank mRNA | AK097787.1 | - | 1 | 201 | ||
BC043218.2 | - | 1 | 400 |

Gene Symbol: LOC100506271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110515.1 | 1 | 399 | |||
GenBank mRNA | AK097787.1 | - | 1 | 201 | ||
BC043218.2 | - | 1 | 400 |