siRNA Details
Gene Symbol: LOC101928748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110950.1 | 3 | 964 | |||
GenBank mRNA | BC034316.1 | - | 3 | 964 |

Gene Symbol: LOC101928748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110950.1 | 3 | 964 | |||
GenBank mRNA | BC034316.1 | - | 3 | 964 |