siRNA Details
Gene Symbol: LOC105377213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135614.1 | 2 | 440 | |||
GenBank mRNA | BX103087.1 | - | 2 | 440 |

Gene Symbol: LOC105377213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135614.1 | 2 | 440 | |||
GenBank mRNA | BX103087.1 | - | 2 | 440 |