siRNA Details
Gene Symbol: LOC101927482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110225.1 | 3 | 977 | |||
NR_110226.1 | 3 | 966 | ||||
GenBank mRNA | AK023739.1 | - | 2 | 966 | ||
BC018675.1 | - | 2 | 856 |

Gene Symbol: LOC101927482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110225.1 | 3 | 977 | |||
NR_110226.1 | 3 | 966 | ||||
GenBank mRNA | AK023739.1 | - | 2 | 966 | ||
BC018675.1 | - | 2 | 856 |