This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LINC01137 SIRNA Assays ›
Gene Symbol:
LINC01137
Gene Name:
long intergenic non-protein coding RNA 1137
Gene Aliases:
-
Chromosome Location:
Chr.1: 37454879 - 37474443 on Build GRCh38
UniGene ID:
Hs.380738
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n548181

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINC01137
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_038842.1 3 494
GenBank mRNA AL162073.1 - 1 49
BM923085.1 - 3 440
DA727774.1 - 2 404
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA HY118888.1 - 3 426 Chr. 1 - 37455762 - 37474375 [-] on Build GRCh38
GenBankmRNA HY161653.1 - 3 372 Chr. 1 - 37455715 - 37474320 [-] on Build GRCh38
GenBankmRNA HY173083.1 - 3 372 Chr. 1 - 37455682 - 37474320 [-] on Build GRCh38
GenBankmRNA HY164243.1 - 3 428 Chr. 1 - 37455732 - 37474374 [-] on Build GRCh38
GenBankmRNA CB989356.1 - 3 450 Chr. 1 - 37455716 - 37474375 [-] on Build GRCh38
GenBankmRNA CX163127.1 - 2 344 Chr. 1 - 37455276 - 37474366 [-] on Build GRCh38
GenBankmRNA HY166869.1 - 3 455 Chr. 1 - 37455749 - 37474402 [-] on Build GRCh38
GenBankmRNA HY077067.1 - 3 444 Chr. 1 - 37455762 - 37474393 [-] on Build GRCh38
GenBankmRNA HY035576.1 - 3 453 Chr. 1 - 37455793 - 37474402 [-] on Build GRCh38
GenBankmRNA CB127415.1 - 3 418 Chr. 1 - 37455783 - 37474367 [-] on Build GRCh38
GenBankmRNA HY159741.1 - 3 411 Chr. 1 - 37455727 - 37474359 [-] on Build GRCh38
GenBankmRNA CR997019.1 - 3 434 Chr. 1 - 37455775 - 37474367 [-] on Build GRCh38
GenBankmRNA CN268437.1 - 3 428 Chr. 1 - 37455623 - 37474367 [-] on Build GRCh38
GenBankmRNA CD110171.1 - 3 489 Chr. 1 - 37455510 - 37474415 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header