This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other THUMPD3-AS1 SIRNA Assays ›
Gene Symbol:
THUMPD3-AS1
Gene Name:
THUMPD3 antisense RNA 1
Gene Aliases:
SETD5-AS1
Chromosome Location:
Chr.3: 9385264 - 9397494 on Build GRCh38
UniGene ID:
Hs.598958
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n546765

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: THUMPD3-AS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027007.3 3 294
NR_132780.1 4 368
NR_132781.1 2 139
GenBank mRNA AK093640.1 - 2 1419
AL110136.1 - 3 290
AW237468.1 - 2 139
BC036698.1 - 2 2744
BC044625.2 - 1 1405
BC052961.1 - 3 2666
BX953112.1 - 4 349
HY142091.1 - 3 223
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BE217739.1 - 2 121 Chr. 3 - 9389511 - 9396605 [-] on Build GRCh38
GenBankmRNA BJ994980.1 - 4 313 Chr. 3 - 9390267 - 9397439 [-] on Build GRCh38
GenBankmRNA AA463541.1 - 3 288 Chr. 3 - 9390388 - 9397483 [-] on Build GRCh38
GenBankmRNA BU159574.1 - 3 273 Chr. 3 - 9389825 - 9397464 [-] on Build GRCh38
GenBankmRNA BM547994.1 - 1 347 Chr. 3 - 9389986 - 9390750 [-] on Build GRCh38
GenBankmRNA DB239124.1 - 4 314 Chr. 3 - 9389699 - 9397439 [-] on Build GRCh38
GenBankmRNA DB087998.1 - 3 240 Chr. 3 - 9389610 - 9397439 [-] on Build GRCh38
GenBankmRNA CN349972.1 - 3 264 Chr. 3 - 9385793 - 9395580 [-] on Build GRCh38
GenBankmRNA CN349971.1 - 2 230 Chr. 3 - 9389492 - 9391828 [-] on Build GRCh38
GenBankmRNA AA442499.1 - 2 206 Chr. 3 - 9390241 - 9391810 [-] on Build GRCh38
GenBankmRNA AA923144.1 - 3 277 Chr. 3 - 9389739 - 9397460 [-] on Build GRCh38
GenBankmRNA BU537437.1 - 3 277 Chr. 3 - 9390067 - 9397474 [-] on Build GRCh38
GenBankmRNA BX344859.2 - 3 266 Chr. 3 - 9389799 - 9397482 [-] on Build GRCh38
GenBankmRNA BG751442.1 - 2 480 Chr. 3 - 9385793 - 9391828 [-] on Build GRCh38
GenBankmRNA BQ428680.1 - 3 267 Chr. 3 - 9390025 - 9397496 [-] on Build GRCh38
GenBankmRNA BI495272.1 - 1 443 Chr. 3 - 9390262 - 9390857 [-] on Build GRCh38
GenBankmRNA AL524186.3 - 3 266 Chr. 3 - 9389625 - 9397482 [-] on Build GRCh38
GenBankmRNA BI756773.1 - 2 242 Chr. 3 - 9389851 - 9391827 [-] on Build GRCh38
GenBankmRNA DB523053.1 - 1 52 Chr. 3 - 9390015 - 9390474 [-] on Build GRCh38
GenBankmRNA AI298618.1 - 3 281 Chr. 3 - 9390288 - 9396605 [-] on Build GRCh38
GenBankmRNA BQ548851.1 - 3 305 Chr. 3 - 9390138 - 9396634 [-] on Build GRCh38
GenBankmRNA AI458342.1 - 3 256 Chr. 3 - 9389727 - 9396580 [-] on Build GRCh38
GenBankmRNA BG943385.1 - 3 282 Chr. 3 - 9385854 - 9398579 [-] on Build GRCh38
GenBankmRNA AA504762.1 - 3 297 Chr. 3 - 9390270 - 9395577 [-] on Build GRCh38
GenBankmRNA HY003086.1 - 3 296 Chr. 3 - 9390164 - 9397494 [-] on Build GRCh38
GenBankmRNA BM712163.1 - 2 126 Chr. 3 - 9389556 - 9391729 [-] on Build GRCh38
GenBankmRNA CR745754.1 - 2 196 Chr. 3 - 9390014 - 9391799 [-] on Build GRCh38
GenBankmRNA CR743394.1 - 2 474 Chr. 3 - 9385898 - 9391823 [-] on Build GRCh38
GenBankmRNA BX423431.2 - 2 470 Chr. 3 - 9389923 - 9391822 [-] on Build GRCh38
GenBankmRNA HY159055.1 - 4 319 Chr. 3 - 9390261 - 9397439 [-] on Build GRCh38
GenBankmRNA AW612073.1 - 3 271 Chr. 3 - 9390382 - 9396607 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header