siRNA Details
Gene Symbol: LOC729867 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123725.1 | 3 | 478 | |||
GenBank mRNA | AK093299.1 | - | 3 | 478 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB030112.1 | - | 3 | 479 | Chr. 1 - 160205972 - 160208869 [-] on Build GRCh38 |
