This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LINC00222 SIRNA Assays ›
Gene Symbol:
LINC00222
Gene Name:
long intergenic non-protein coding RNA 222
Gene Aliases:
C6orf181, NCRNA00222, dJ354J5.2
Chromosome Location:
Chr.6: 108751654 - 108769942 on Build GRCh38
UniGene ID:
Hs.520310
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n544715

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINC00222
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033376.1
2 244
GenBank mRNA AY927443.1 - LIT3472
AY927443
2 245
AY927444.1 - LIT3473
AY927444
2 242
AY927445.1 -
2 212
AY927446.1 - LIT3474
AY927446
2 245
AY927447.1 - LIT3475
AY927447
2 207
AY927448.1 - LIT3476
AY927448
2 86
AY927449.1 - LIT3477
AY927449
2 80

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0