siRNA Details
Gene Symbol: LOC105377763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134259.1 | 3 | 582 | |||
GenBank mRNA | DA957262.1 | - | 3 | 439 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI523725.1 | - | 3 | 454 | Chr. 5 - 179658236 - 179664432 [+] on Build GRCh38 |
