This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LINC01028 SIRNA Assays ›
Gene Symbol:
LINC01028
Gene Name:
long intergenic non-protein coding RNA 1028
Gene Aliases:
TCONS_00025127
Chromosome Location:
Chr.17: 70051271 - 70068095 on Build GRCh38
UniGene ID:
Hs.639899
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n544063

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINC01028
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_104129.1 3 456
NR_104130.1 2 334
GenBank mRNA BX647864.1 - 2 328
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BG189623.1 - 1 199 Chr. 17 - 70067183 - 70067827 [+] on Build GRCh38
GenBankmRNA AL691655.1 - 2 328 Chr. 17 - 70051277 - 70067786 [+] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0