siRNA Details
Gene Symbol: HCP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040662.1 | 2 | 134 | |||
GenBank mRNA | AK093953.1 | - | 2 | 129 | ||
AK290875.1 | - | 2 | 129 | |||
BC106759.2 | - | 2 | 80 | 40024782 | ||
BC113853.1 | - | 1 | 171 | |||
BC114462.1 | - | 2 | 80 | |||
BX640979.1 | - | 2 | 132 | |||
DA501560.1 | - | 2 | 135 | |||
L06175.1 | - | 2 | 123 |
