This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC100507073 SIRNA Assays ›
Gene Symbol:
LOC100507073
Gene Name:
uncharacterized LOC100507073
Gene Aliases:
-
Chromosome Location:
Chr.2: 66574030 - 66695588 on Build GRCh38
UniGene ID:
Hs.667940
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n541196

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC100507073
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_110156.1 5 348
GenBank mRNA CN360491.1 - 5 348
DA606892.1 - 2 130
DA610966.1 - 2 145
DA614164.1 - 2 153
DA614343.1 - 2 156
DA616181.1 - 2 153
DA618279.1 - 2 138
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA610069.1 - 2 134 Chr. 2 - 66691435 - 66695583 [+] on Build GRCh38
GenBankmRNA DA612877.1 - 1 279 Chr. 2 - 66691416 - 66695488 [+] on Build GRCh38
GenBankmRNA DA604628.1 - 2 153 Chr. 2 - 66691416 - 66695488 [+] on Build GRCh38
GenBankmRNA DA610500.1 - 1 258 Chr. 2 - 66691435 - 66695465 [+] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0