This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 1700041C23RIK SIRNA Assays ›
Gene Symbol:
1700041C23Rik
Gene Name:
RIKEN cDNA 1700041C23 gene
Gene Aliases:
-
Chromosome Location:
Chr.9: 57175275 - 57186108 on Build GRCm38
UniGene ID:
Mm.234187
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n522892

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1700041C23Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033784.1 4 251
GenBank mRNA AK006669.1 - 4 253
AK007035.1 - 4 252
AK018934.1 - 4 241
BY707168.1 - 4 241
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CN838902.1 - 242 Chr. - - [-] on Build
GenBankmRNA CA465030.1 - 4 253 Chr. 9 - 57175275 - 57186108 [+] on Build GRCm38
GenBankmRNA CA465155.1 - 243 Chr. - - [-] on Build
GenBankmRNA BY707103.1 - 4 252 Chr. 9 - 57175275 - 57186107 [+] on Build GRCm38
GenBankmRNA BY706656.1 - 4 252 Chr. 9 - 57175275 - 57186106 [+] on Build GRCm38
GenBankmRNA BU961366.1 - 211 Chr. - - [-] on Build

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0