This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 0610031O16RIK SIRNA Assays ›
Gene Symbol:
0610031O16Rik
Gene Name:
RIKEN cDNA 0610031O16 gene
Gene Aliases:
-
Chromosome Location:
Chr.3: 138210717 - 138239663 on Build GRCm38
UniGene ID:
Mm.441136
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n522613

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 0610031O16Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_045760.1 1 44
GenBank mRNA AK002733.1 - 1 44
BC044770.1 - 119
BC046436.1 - 119
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI144451.1 - 2 120 Chr. 3 - 138229834 - 138240405 [-] on Build GRCm38
GenBankmRNA BY703239.1 - 1 44 Chr. 3 - 138210716 - 138239663 [-] on Build GRCm38
GenBankmRNA BB598260.2 - 1 61 Chr. 3 - 138226560 - 138239679 [-] on Build GRCm38
GenBankmRNA AA237879.1 - 1 44 Chr. 3 - 138217793 - 138239663 [-] on Build GRCm38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0