This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNHG6 SIRNA Assays ›
Gene Symbol:
Snhg6
Gene Name:
small nucleolar RNA host gene 6
Gene Aliases:
1110008H02Rik
Chromosome Location:
Chr.1: 9942025 - 9944118 on Build GRCm38
UniGene ID:
Mm.28311
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n522514

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Snhg6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024067.2 4 290
GenBank mRNA AK003573.1 - 4 291
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CJ320246.1 - 2 328 Chr. 1 - 9942026 - 9942683 [-] on Build GRCm38
GenBankmRNA BY788907.1 - 4 289 Chr. 1 - 9942074 - 9944117 [-] on Build GRCm38
GenBankmRNA AA015539.1 - 4 250 Chr. 1 - 9942042 - 9944079 [-] on Build GRCm38
GenBankmRNA AA023387.1 - 4 252 Chr. 1 - 9942023 - 9944083 [-] on Build GRCm38
GenBankmRNA BY604390.1 - 4 295 Chr. 1 - 9942019 - 9944118 [-] on Build GRCm38
GenBankmRNA AV115890.2 - 469 Chr. - - [-] on Build
GenBankmRNA AA250387.1 - 4 267 Chr. 1 - 9942054 - 9944087 [-] on Build GRCm38
GenBankmRNA BI109518.1 - 4 273 Chr. 1 - 9942023 - 9944099 [-] on Build GRCm38
GenBankmRNA AV485517.1 - 4 277 Chr. 1 - 9942023 - 9944104 [-] on Build GRCm38
GenBankmRNA BY345355.1 - 4 293 Chr. 1 - 9942107 - 9944121 [-] on Build GRCm38
GenBankmRNA BY111725.1 - 4 289 Chr. 1 - 9942091 - 9944117 [-] on Build GRCm38
GenBankmRNA AA097989.1 - 4 260 Chr. 1 - 9942025 - 9944095 [-] on Build GRCm38
GenBankmRNA AV468093.1 - 4 250 Chr. 1 - 9942028 - 9944077 [-] on Build GRCm38
GenBankmRNA BE134757.1 - 4 270 Chr. 1 - 9942023 - 9944098 [-] on Build GRCm38
GenBankmRNA W35693.1 - 219 Chr. - - [-] on Build
GenBankmRNA BX523010.1 - 4 280 Chr. 1 - 9942023 - 9944107 [-] on Build GRCm38
GenBankmRNA AA267148.1 - 3 128 Chr. 1 - 9942023 - 9942654 [-] on Build GRCm38
GenBankmRNA W18398.1 - 259 Chr. - - [-] on Build
GenBankmRNA AA023086.1 - 4 279 Chr. 1 - 9942024 - 9944105 [-] on Build GRCm38
GenBankmRNA AA119805.1 - 4 265 Chr. 1 - 9942023 - 9944098 [-] on Build GRCm38
GenBankmRNA AA940467.1 - 4 269 Chr. 1 - 9941959 - 9944090 [-] on Build GRCm38
GenBankmRNA W40695.1 - 218 Chr. - - [-] on Build
GenBankmRNA BX516798.1 - 4 280 Chr. 1 - 9942023 - 9944105 [-] on Build GRCm38
GenBankmRNA AA002823.1 - 4 273 Chr. 1 - 9942023 - 9944098 [-] on Build GRCm38
GenBankmRNA BY320914.1 - 4 318 Chr. 1 - 9942114 - 9944117 [-] on Build GRCm38
GenBankmRNA AA289428.1 - 3 175 Chr. 1 - 9942025 - 9942695 [-] on Build GRCm38
GenBankmRNA W54219.1 - 215 Chr. - - [-] on Build
GenBankmRNA BY345466.1 - 4 293 Chr. 1 - 9942116 - 9944121 [-] on Build GRCm38
GenBankmRNA BY243424.1 - 4 290 Chr. 1 - 9942019 - 9944117 [-] on Build GRCm38
GenBankmRNA W11261.1 - 273 Chr. - - [-] on Build
GenBankmRNA BY034531.1 - 4 290 Chr. 1 - 9942060 - 9944117 [-] on Build GRCm38
GenBankmRNA W98878.1 - 268 Chr. - - [-] on Build
GenBankmRNA AA146116.1 - 4 210 Chr. 1 - 9942033 - 9944038 [-] on Build GRCm38
GenBankmRNA AW988981.1 - 4 270 Chr. 1 - 9942023 - 9944098 [-] on Build GRCm38
GenBankmRNA AA098159.1 - 4 267 Chr. 1 - 9942037 - 9944097 [-] on Build GRCm38
GenBankmRNA AA155393.1 - 4 215 Chr. 1 - 9942025 - 9944043 [-] on Build GRCm38
GenBankmRNA BY703829.1 - 4 289 Chr. 1 - 9942025 - 9944117 [-] on Build GRCm38
GenBankmRNA BF715116.1 - 4 277 Chr. 1 - 9942023 - 9944105 [-] on Build GRCm38
GenBankmRNA AA124208.1 - 4 274 Chr. 1 - 9942058 - 9944104 [-] on Build GRCm38
GenBankmRNA AA240411.1 - 3 128 Chr. 1 - 9942039 - 9942654 [-] on Build GRCm38
GenBankmRNA BY711012.1 - 4 289 Chr. 1 - 9941960 - 9944117 [-] on Build GRCm38
GenBankmRNA W54183.1 - 272 Chr. - - [-] on Build
GenBankmRNA AV490092.1 - 4 254 Chr. 1 - 9942038 - 9944081 [-] on Build GRCm38
GenBankmRNA BY344080.1 - 4 294 Chr. 1 - 9942094 - 9944121 [-] on Build GRCm38
GenBankmRNA AV115922.2 - 4 311 Chr. 1 - 9941973 - 9944118 [-] on Build GRCm38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-6889b944d9-flx6z:80/100.66.128.142:80.
git-commit: df865406a845caddbd23a531a4b79e504db49c75
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header