This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 4933404O12RIK SIRNA Assays ›
Gene Symbol:
4933404O12Rik
Gene Name:
RIKEN cDNA 4933404O12 gene
Gene Aliases:
AW494809
Chromosome Location:
Chr.5: 136919146 - 136937109 on Build GRCm38
UniGene ID:
Mm.135764
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n522407

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4933404O12Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015555.1 3 427
GenBank mRNA AK006082.1 - 3 186
AK016656.1 - 3 187
AK085506.1 - 3 428
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CK634823.1 - 3 393 Chr. 5 - 136919180 - 136933942 [+] on Build GRCm38
GenBankmRNA BY285462.1 - 3 183 Chr. 5 - 136919391 - 136933793 [+] on Build GRCm38
GenBankmRNA BB641313.1 - 3 408 Chr. 5 - 136919165 - 136933827 [+] on Build GRCm38
GenBankmRNA BY716344.1 - 3 187 Chr. 5 - 136919386 - 136934224 [+] on Build GRCm38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0