siRNA Details
Gene Symbol: AI115009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040386.1 | 4 | 1778 | |||
GenBank mRNA | AK043786.1 | - | 4 | 1781 |

Gene Symbol: AI115009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040386.1 | 4 | 1778 | |||
GenBank mRNA | AK043786.1 | - | 4 | 1781 |