This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MIRG SIRNA Assays ›
Gene Symbol:
Mirg
Gene Name:
miRNA containing gene
Gene Aliases:
2810474H01Rik, AI852992, Gm2928, Meg9, mir-410
Chromosome Location:
Chr.12: 109734981 - 109749458 on Build GRCm38
UniGene ID:
Mm.260543
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n521703

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mirg
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028265.1
8 991
GenBank mRNA AF498297.1 -
3 263
AF498298.1 -
3 263
AJ517767.1 - LIT1749
Mirg
8 991
AK013406.1 -
3 286
AK081334.1 -
1 1247
EU434920.1 -
2876
EU616812.1 -
3513
EU616813.1 -
3488
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CB590423.1 - 374 Chr. - - [-] on Build
GenBankmRNA AA058156.1 - 3 266 Chr. 12 - 109743808 - 109749310 [+] on Build GRCm38
GenBankmRNA AA024068.1 - 3 247 Chr. 12 - 109743825 - 109749368 [+] on Build GRCm38
GenBankmRNA CK129636.1 - 4 326 Chr. 12 - 109743801 - 109749460 [+] on Build GRCm38
GenBankmRNA W99879.1 - 244 Chr. - - [-] on Build
GenBankmRNA AV464287.1 - 3 250 Chr. 12 - 109743822 - 109749341 [+] on Build GRCm38
GenBankmRNA BG270635.1 - 3 263 Chr. 12 - 109743809 - 109749460 [+] on Build GRCm38
GenBankmRNA CJ282916.1 - 2 195 Chr. 12 - 109748154 - 109749454 [+] on Build GRCm38
GenBankmRNA BY712398.1 - 3 286 Chr. 12 - 109743784 - 109749455 [+] on Build GRCm38
GenBankmRNA W65931.1 - 3 242 Chr. 12 - 109743830 - 109749375 [+] on Build GRCm38
GenBankmRNA CF586443.1 - 3 285 Chr. 12 - 109743804 - 109749460 [+] on Build GRCm38
GenBankmRNA BM894903.1 - 2 342 Chr. 12 - 109747998 - 109749320 [+] on Build GRCm38
GenBankmRNA BY608086.1 - 2 90 Chr. 12 - 109748259 - 109749454 [+] on Build GRCm38
GenBankmRNA CF586254.1 - 3 275 Chr. 12 - 109743804 - 109749460 [+] on Build GRCm38
GenBankmRNA AV500425.1 - 2 200 Chr. 12 - 109748157 - 109749347 [+] on Build GRCm38
GenBankmRNA BQ086866.1 - 2 353 Chr. 12 - 109747987 - 109749355 [+] on Build GRCm38
GenBankmRNA AA024074.1 - 3 247 Chr. 12 - 109743825 - 109749369 [+] on Build GRCm38
GenBankmRNA W50730.1 - 3 239 Chr. 12 - 109743832 - 109749358 [+] on Build GRCm38
GenBankmRNA CB203581.1 - 3 254 Chr. 12 - 109743817 - 109749460 [+] on Build GRCm38
GenBankmRNA W50748.1 - 146 Chr. - - [-] on Build
GenBankmRNA W85008.1 - 239 Chr. - - [-] on Build

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0