siRNA Details
Gene Symbol: 4933416M07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045840.1 | 1 | 52 | |||
GenBank mRNA | AK016835.1 | - | 1 | 52 |

Gene Symbol: 4933416M07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045840.1 | 1 | 52 | |||
GenBank mRNA | AK016835.1 | - | 1 | 52 |