siRNA Details
Gene Symbol: 1110020A21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027930.1 | 2 | 595 | |||
GenBank mRNA | AK011222.1 | - | 2 | 172 | ||
BI737205.1 | - | 2 | 389 |

Gene Symbol: 1110020A21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027930.1 | 2 | 595 | |||
GenBank mRNA | AK011222.1 | - | 2 | 172 | ||
BI737205.1 | - | 2 | 389 |